Would you like to log in again?
Cubis® II Semi-Micro Balances
Supplier: Sartorius
Cubis® II
MCA225P3S00DQP99 MCA225P3S00DQP99 MCA225P3S00DMDS MCA225P3S00DQP99ION MCA225P3S00DION MCA226S3S00DQP99ION MCE125S3S00D MCA225S3S00DHWL MCA226S3S00DION MCE225P3S00DION MCA226S3S00DQP99 MCE225P3S00DION MCA226S3S00DQP99 MCA226S3S00DQP99ION MCE225S3S00D MCE225P3S00D MCA225S3S00DQP99ION MCA226S3S00DION MCA225S3S00DQP99 MCA125S3S00D MCE225P3S00DHWL MCA125S3S00DION MCE225P3S00D MCA225S3S00DQP99ION MCA225S3S00D MCA225S3S00DHWL MCA125S3S00DMDS MCE125S3S00DMDS MCA225S3S00DMDS MCA226S3S00DMDS MCA125S3S00DHWL MCA225S3S00DION MCE225S3S00D MCA225P3S00DQP99HWL MCA125S3S00D MCA226S3S00D MCA125S3S00DMDS MCA226S3S00DQP99MDS MCA225S3S00D MCA225P3S00DHWL MCE125S3S00D MCE225S3S00DMDS MCE225S3S00DION MCA225P3S00DHWL MCA225P3S00DQP99HWL MCE125S3S00DMDS MCA225S3S00DQP99 MCA226S3S00DHWL MCE225P3S00DHWL MCE125S3S00DION MCA225P3S00DION MCE225S3S00DION MCA225S3S00DQP99MDS MCA225P3S00DQP99ION MCA225P3S00DQP99MDS MCA125S3S00DHWL MCE225P3S00DMDS MCA226S3S00DMDS MCA226S3S00DQP99MDS MCE225P3S00DMDS MCA225P3S00DQP99MDS MCA226S3S00DHWL MCA225P3S00D MCE125S3S00DION MCE225S3S00DMDS MCA225S3S00DION MCA225S3S00DMDS MCA225P3S00DMDS MCA225P3S00D MCA225S3S00DQP99MDS MCA226S3S00D MCA125S3S00DION
77476-538EA 29580 USD InStock
CA77476-504 CA77476-548 77476-538 CA77476-506 CA77476-508 77476-540 77476-542 CA77476-550 77476-544 77476-500 CA77476-552 77476-502 77476-546 CA77476-554 CA77476-510 77476-504 77476-548 CA77476-512 CA77476-556 77476-506 CA77476-514 CA77476-558 77476-508 CA77476-516 CA77476-518 77476-550 77476-552 CA77476-520 77476-554 77476-510 CA77476-522 CA77476-488 77476-512 77476-556 CA77476-524 77476-514 77476-558 CA77476-526 77476-516 CA77476-528 77476-518 CA77476-490 CA77476-492 CA77476-494 77476-520 CA77476-496 77476-522 CA77476-498 CA77476-530 77476-488 77476-524 CA77476-532 77476-526 CA77476-534 77476-528 CA77476-536 CA77476-538 77476-490 77476-492 77476-494 77476-496 CA77476-540 77476-498 77476-530 CA77476-542 77476-532 CA77476-500 CA77476-544 77476-534 CA77476-546 CA77476-502 77476-536
Cubis® II Semi-Micro Balances
Balances and Scales
Semi-micro balances come with a readability of 0.01 mg or 10 µg and a maximum weighing capacity of up to 220 g.
  • Superior weighing performance: Fast measurement time and accurate weighing results are guaranteed thanks to next generation monolithic weighing system and the engineering solution compensating for environmental changes
  • No more issues with charged samples: Glass draft shields coated with conductive layer prevent outer electrostatic effects and the built-in ionizer effectively eliminates electrostatic charges from samples
  • Cleaning process guidance: Intuitive process for daily and advanced cleaning is now available in all Cubis® II MCA. The cleaning QApp provides visual guidance, information about chemical compatibility as well as electronical tracking of these events
  • Hardware upgradeability: Hardware features like motorized draft shield function or built-in ionizer can be activated post-purchase. An automated inner draft shield is available as a klick-in accessory which is recommended for use under challenging environmental conditions (e.g. in drafty environments)
  • Compliance and Data Integrity: The end-to-end data integrity, technical controls for 21 CFR Part 11 compliance, integrated audit-trail and state-of-the-art user management ensure laboratories can meet regulatory requirements. These features are directly available on the Cubis® II balances, without the need of additional software
  • Balances Fleet Management: The Ingenix Suite is a flexible, open solution that works with or without an ELN/LIM system. It offers unlimited connections to easily manage the entire Cubis® II MCA lab balance fleet across all labs within the same network
  • Ease of use: The teaching function with learning capability of the motorized draft shield, guided workflows for many different applications (QApps), automated motorized leveling and automatic internal adjustment (isoCAL) help with easy and error-free operation of the balance
Semi-micro balances are used for weighing small sample amounts down to the µg-range. To meet the requirements of your specific application, specially designed weighing pans, sample holders and software applications are available to simplify your weighing processes and laboratory workflows.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty