Would you like to log in again?
Heratherm™ Environmental Chambers
Supplier: Thermo Scientific
Heratherm™
EIS750 ECS750 ELG750 ESG750 EIG750 ECG750 ELS750 ESS750
77486-596EA 26400.5 USD InStock
77486-596 77486-608 77486-606 77486-604 77486-602 77486-610 77486-600 77486-598
Heratherm™ Environmental Chambers
Testing Chambers Environmental Test Chambers
Heratherm Environmental chambers offer exceptional environmental control in a hassle-free chamber that is easy to program and use.
  • Reduce your environmental footprint while achieving outstanding results with Heratherm Environmental chamber's decreased water and energy consumption and Peltier technology
  • Water-efficient humidification system with integrated water tank and optional clean water connection
  • Peltier technology supports temperature stability and uniformity with lower energy usage compared to traditional compressors
  • Vertical low-energy LED lighting includes UV light and programmable light spectrum and day/night cycles
  • Four models available offering various controlled climates including humidity, temperature, light and UV light for testing, and CO₂ incubation
  • Security features including overtemperature alarm and switch off features, door alarms, restricted protocol programming, various alerts, and manual/automated door locking
  • Ease-of-use features including dual access ports for maximum flexibility and rounded corners for easy cleaning
Sleek in appearance, these chambers blend aesthetics and precise environmental controls with cutting-edge technology. Thermo Scientific™ Heratherm™ Environmental chambers offer reliable and consistent performance supported by data capture and validation for 21 CFR Part 11. They are excellent for use in controlled environments with flexible configurations to meet the demands of academic and industrial workflows.
Certifications: CE, UL.
Ordering information: Accessories sold separately.
Delivery information: Inside delivery is recommended due to size of chamber.
Packaging: The bottles are packaged in shrinked-wrapped modules.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty