Would you like to log in again?
SAFECAP® Capillary Tubes, Self-Sealing Mylar® Wrapped
Supplier: Drummond
P75-2000M R60-2000M H75-2000M
77497-226BX 416 USD InStock
77497-226 77497-228 77497-222
SAFECAP® Capillary Tubes, Self-Sealing Mylar® Wrapped
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Blood Sample Collection/Preparation
SAFECAP® is a single-use, Triple Mylar® wrapped capillary tube with a super absorbent plug that automatically seals on contact with blood.
  • Unique
  • Self sealing capillary tubes
  • Mylar® wrap minimizes risk of exposure to HIV, hepatitis, and other bloodborne pathogens
  • Case of 20 vials contains 100 tubes each
The Mylar® wrap significantly reduces exposure to potentially life-threatening blood-borne pathogens and glass fragments, while the self-sealing plug eliminates manually sealing with clay and reduces centrifuge blowouts.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty