Would you like to log in again?
Safepette® Dispenser
Supplier: Drummond
Safepette®
SD2050
77497-230BX 320 USD InStock
77497-230
Safepette® Dispenser
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Blood Sample Collection/Preparation
After centrifugation, a Safecap® tube is loaded into the Safepette® dispenser. The plunger is then depressed to move the self-sealing plug, which acts as a piston.
This eliminates the hazardous practice of scoring and snapping blood-filled tubes and the associated risk of exposure to potentially life-threatening blood-borne pathogens.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty