Would you like to log in again?
Horse Blood Agar, 5%
Supplier: Hardy Diagnostics
A149 A149
77497-252PK 210.77 USD InStock
77497-252 CA77497-252
Horse Blood Agar, 5%
Media for Microbiology Prepared Media for Microbiology
Contains 5% defibrinated horse blood for the cultivation of Haemophilus spp.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty