Would you like to log in again?
Urine Quad Plate
Supplier: Hardy Diagnostics
J71 J71
77497-256PK 105.8 USD InStock
77497-256 CA77497-256
Urine Quad Plate
Media for Microbiology Prepared Media for Microbiology
EMB Agar formulations are recommended for use as selective and differential media for the isolation of Gram-negative bacilli (including coliform organisms and enteric pathogens) from clinical and nonclinical specimens.
  • Optically clear polystyrene
  • Heavy-Duty
  • Stacking rings
  • Breathable packaging
XLD Agar is recommended for use as a selective and differential medium for the isolation of Gram-negative enteric pathogens from fecal specimens or from food and other samples.
Citrate Agar is recommended for use in the differentiation of Gram-negative enteric bacilli based on citrate utilization.
Blood Agar products are recommended for use as general purpose growth media for the isolation, cultivation, and differentiation of a wide variety of microorganisms.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty