Would you like to log in again?
ClikSeal™ Specimen Cup, with Lid, Sterile
Supplier: Hardy Diagnostics
ClikSeal™
PC4064200S PC4064200S
77497-260PK 264.14 USD InStock
77497-260 CA77497-260
ClikSeal™ Specimen Cup, with Lid, Sterile
Sample Containers General Purpose Containers
ClikSeal™ represents a new advance in container technology. The patented lid is designed to securely lock in the sample. It even lets you know when the seal has been made by sounding an audible CLICK when the cap is screwed into the secure position. When you hear the click, you can be confident that your sample is safely contained in the cup and will remain that way throughout transport to the lab!
  • The lid features a nesting design so that the cups can be easily stacked
  • Leak-Proof containers are 95 kPa compliant
  • ClikSeals™ are constructed of tough polypropylene with thick walls making them virtually indestructible
  • Easy to read paper label is attached for easy sample identification. Graduation marks on the side allow for volume measurement
Widely use in breweries, wineries and distilleries, cannabis QA testing, clinical, cosmetics, environmental, food safety, pharmaceutical manufacturing industries.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty