Would you like to log in again?
ONPG Disks
Supplier: Hardy Diagnostics
Z7321 Z7321
77497-262PK 149.34 USD InStock
77497-262 CA77497-262
ONPG Disks
Food and Beverage Test Systems
ONPG differentiation disk is used to differentiate members of the enterobacteriaceae and other microorganisms based on beta-galactosidase activity.
Ordering information: Each cartridge contains 50 disks that are spring loaded for smooth and reliable dispensing.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty