Would you like to log in again?
HardyDisk™ AST Cephalothin, CF30
Supplier: Hardy Diagnostics
HardyDisks™
Z8335 Z8335
77497-266PK 155.27 USD InStock
CA77497-266 77497-266
HardyDisk™ AST Cephalothin, CF30
Microbiology Test Systems Antibiotic Tests
HardyDisk™ AST's are impregnated paper disks used for Antimicrobial Susceptibility Testing (AST) also known as disk diffusion or Kirby-Bauer testing.
Ordering information: Each cartridge contains 50 disks that are spring loaded for smooth and reliable dispensing.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty