Would you like to log in again?
HardyDisk™ AST Imipenem/Relebactam, IMR10/25
Supplier: Hardy Diagnostics
HardyDisks™
Z9441 Z9441
77497-268PK 43.67 USD InStock
77497-268 CA77497-268
HardyDisk™ AST Imipenem/Relebactam, IMR10/25
Microbiology Test Systems Antibiotic Tests
HardyDisk™ AST's are impregnated paper disks used for Antimicrobial Susceptibility Testing (AST); also known as disk diffusion or Kirby-Bauer testing.
  • Hardy Diagnostics offers a complete line of all commonly used antibiotics
  • Compatible with the BBL dispensers
  • Improved cartridge design helps to prevent jamming and tearing
  • HardyDisk™ AST cartridges are packaged in an all new canister that features a desiccant built into the lid
  • Kirby-Bauer accessories available: Mueller Hinton Agar, McFarland Standard, calipers, and much more
  • The last disk in each cartridge is clearly marked with dark lines around its edge to signify the need for refill
Ordering information: Each cartridge contains 50 disks that are spring loaded for smooth and reliable dispensing.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty