Would you like to log in again?
HardyDisk™ Rezafungin, 5 µg, (RZF5)
Supplier: Hardy Diagnostics
HardyDisks™
Z9471 Z9471
77497-274PK 39.67 USD InStock
77497-274 CA77497-274
HardyDisk™ Rezafungin, 5 µg, (RZF5)
Microbiology Test Systems Antibiotic Tests
Rezafungin is a new antifungal agent that combats invasive fungal infections. This marks the first new treatment option approved for individuals with candidemia and invasive candidiasis in over ten years.
Ordering information: Each cartridge contains 50 disks that are spring loaded for smooth and reliable dispensing.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty