Would you like to log in again?
BradyPrinter i5300 Heat Shrink Label Printer with Wi-Fi and Product and Wire ID Software
Supplier: Brady
151294
77506-646EA 3755.3 USD InStock
77506-646
BradyPrinter i5300 Heat Shrink Label Printer with Wi-Fi and Product and Wire ID Software
Printers Label Printers
Get fast and efficient precision printing for high-mix/high-volume labeling. Printer and software allow for quick label set up and no-waste material changeovers.
  • 600 dpi; prints small fonts with precision
  • Requires no calibration and no heat and speed setting adjustments
  • 20-second material changeovers, 0.20 to 4" label widths
  • Pauses, resumes and stores print jobs on printer in an adjustable queue with overwrite protection
  • 7" color touchscreen and built-in auto cutter
  • Solid construction with an all-metal frame
  • 1-click label part recognition and setup with Brady Workstation Product and Wire ID Software Suite
  • Intuitive and easy to operate
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty