Would you like to log in again?
Low-Carbon Centrifuge Tubes, 15 and 50 ml, PP
Supplier: Polycarbin
PC01-01-01G PC02-02-01G PC02-01-01B PC02-01-01G PC01-02-01G PC02-02-01B PC01-02-01B PC01-01-01B
77321-866CS 152.25 USD InStock
77321-866 77506-652 77506-654 77321-864 76635-513 77506-650 77506-648 76635-503
Low-Carbon Centrifuge Tubes, 15 and 50 ml, PP
Tubes Centrifuge Tubes
Polycarbin's centrifuge tubes allow labs to reduce reliance on fossil fuels, as they are made using 20% post-consumer recycled content by weight. These centrifuge tubes combine high performance with a significantly reduced carbon footprint.
  • Lower carbon impact compared to conventional concial tubes
  • Polypropylene (PP) tube, HDPE screw cap (leak-resistant)
  • Sterile, Dnase-, Rnase-, and pyrogen-free
  • Racked and bagged configurations with color-coded caps
  • Sturdy, easy to recycle cardboard rack
Polycarbin's low-carbon laboratory products are made for the sustainability-minded scientist. These centrifuge tubes deliver the expected quality and performance, while allowing the laboratory to reduce its dependence on 100% fossil fuel based plastics. By using low-carbon laboratory products, users can support a less-wasteful, low-carbon supply chain.
Certifications: Certified DNase-, RNase-, and pyrogen-free. Manufactured in an ISO 9001 compliant facility.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty