Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Distribution Pump

Supplier: Avidity Sciences
2300-3830-001
77506-662EA 921.39 USD InStock
77506-662
Distribution Pump
Pumps Drum/Transfer Pumps
The distribution pumps draw treated water from the storage tank and fill an air-pressurized water tank. Two distribution pumps are included to allow for automatic backup operation if one pump should fail. Normally, the pumps alternately operate every two hours to prevent stagnant conditions in either pump.

Distribution pump temperature sensor: A temperature sensor is connected to both distribution pumps to monitor the temperature of the water leaving the pumps. The temperature of the water is then transmitted back to the RO controller.

Distribution pump pressure sensor: A pressure sensor is connected to the distribution pumps to monitor the pressure in the pressure tank. The pressure value is then transmitted back to the RO controller.

Feed water source: Tap
Water quality: <1400 µS, 1 to 15 MΩ-cm, 18.2 MΩ-cm

Water technology type: Type 3, Type 2
Industry: Life Science, Vivarium

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty