Would you like to log in again?
BioXp® Select DNA Cloning Gibson Assembly Kits
Supplier: Telesis Bio
BioXp®
BX-SEL-CLNGA-08 BX-SEL-CLNGA-24
77506-810EA 7.6 USD InStock
77506-810 77506-808
BioXp® Select DNA Cloning Gibson Assembly Kits
Nucleic Acid Reagents Cloning Kits and Ligation
The Select DNA cloning Gibson assembly kits can generate Gibson clones starting with fragments and vector supplied by the end user.
  • Accepted input: Telesis Bio DNA fragments, IDT gBlocks™, Twist Biosciences fragments
  • Supports user-provided linearized vector
  • Length: 0.3 to 6.0 kb
  • Compatible with BioXp® 3250
The Select DNA cloning Gibson assembly kits can generate Gibson clones starting with fragments and vector supplied by the end user.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty