Would you like to log in again?
DuraTube™ and Cap Assemblies, Non-Sterile
Supplier: Scientific Specialties
2641-0B
77506-812CS 288.61 USD InStock
77506-812
DuraTube™ and Cap Assemblies, Non-Sterile
Tubes Microcentrifuge Tubes
Homogenization calls for a tube that is built to withstand the rigors of lysing applications. So when you need to be sure about the tube you select for lysing and bead beating, choose the brand that has the experience, innovation, and reliability of an industry leader: SureSeal DuraTubes from SSIbio.
  • Manufactured from a proprietary resin that is specially-formulated for bead-beating applications
  • Shatter-resistant, they are up to the task
  • Secure O-ring seal ensures a tight seal
  • Ultra-pure polypropylene meets FDA 21 CFR 177.1520 & USP Class VI
  • Graduated every 100 µl for easy volume estimation
  • Self-standing skirted base
Certified free of detectable RNase, DNase, DNA, PCR inhibitors, and tested pyrogen-free.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty