Would you like to log in again?
Cold Resistant Gloves, TempIce 700
Supplier: Mapa Professionnel
34700010 34700019 34700017 34700018
77506-876CS 510 USD InStock
77506-876 77506-878 77506-872 77506-874
Cold Resistant Gloves, TempIce 700
Gloves
These knitted gloves with insulating lining and nitrile grip and proof coating on palm and fingertips provide protection in cold environment down to −10 °C while maintaining comfort, dexterity and flexibility.
  • Five times as durable as comparable standard gloves on the market
  • Excellent grip when handling wet parts
  • Stay dry when working in wet conditions
  • Washable (for five cleaning cycles)
  • Suitable for frozen, dry and some fatty foods use according to the european directive CEE 1935/2004
Typical applications include: Order preparation, maintenance in wet or cold environments, construction materials handling, waste collection, catering services, public park maintenance, maintaining roads.
Certifications: EN 388:2016 (3222X), EN 511 (02X); ISO 13997: N (690g); Cat. II.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty