Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

FluidX 1.6 ml Tri-coded Tubes, 48-format, External Thread

Supplier: Azenta Life Sciences
65-7653 65-7652 65-7650 65-7651
77506-898CS 927.5 USD InStock
77506-898 77506-900 77506-896 77506-902
FluidX 1.6 ml Tri-coded Tubes, 48-format, External Thread
Vials
FluidX 1.6 ml maximum recovery tube is ideally suited for automated workflows using liquid handling, where more sample can be retrieved due to the unique shaped bottom.

  • Sample identification from 1D, 2D and human readable code
  • Capability to work with automation, designed for liquid handlers
  • Suitable for cryogenic storage
  • Made from high-quality virgin PP
  • Available in 48-well format SBS racks or bulk
  • 480 tubes per case or 10 racks per case

Tri-coded tubes offer unequaled sample audit traceability, enabling sample tracking and data sharing between multiple users, labs, locations and automation capabilities.

Suitable for use with liquid handling automation, whether reformatting, cherry picking, aliquoting an/or normalization. The 1.6 ml maximum recovery tube offers unequaled sample audit traceability, enabling sample tracking and data sharing between multiple users, labs, locations and automation capabilities.

Designed and developed with broad compatibility in mind, the tubes perform without compromise in conjunction with automated barcode reading, capping and sample management systems from Azenta Life Sciences and all other industry-recognized manufacturers.

Storage temperature: Minimum temperature (with screw cap): −196 °C (gas phase liquid nitrogen)

Applications: (For research use only) Liquid handling workflows, biobanking, cryogenic storage, cancer research, cellular and molecular biology, drug discovery, compound libraries, reagent storage, CDMOs.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty