Would you like to log in again?
Sample Vials for Vi-CELL BLU Analyzers
Supplier: Beckman Coulter
C99606 C99606
77507-136EA 221.66 USD InStock
77507-136 CA77507-136
Sample Vials for Vi-CELL BLU Analyzers
Vials
Bag contains 500 sample vials for use with the Vi-CELL BLU Analyzer.
  • Minimum volume required in sample vial is 200 µm
  • In FAST mode, a minimum volume of 170 µm is required in the sample vial
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty