Would you like to log in again?
Comparator XPR10U Balance
Supplier: Mettler-Toledo
30756546
77507-138EA 68000 USD InStock
77507-138
Comparator XPR10U Balance
Balances and Scales
Comparator and higher performance balance comes with 10.1 g capacity and 0.0001 mg readability.
  • Highly accurate mass determination
  • Minimized weighing errors
  • Secure data transfer
  • Highly ergonomic operation
  • Automated draftshield
Accurate mass determination:
METTLER TOLEDO mass comparators and higher performance balances guarantee top measurement performance and full traceability of weighing results.
Minimized weighing errors:
Error-free operation through user guidance on the color touchscreen for a smooth and hands-free process.
Secure data transfer:
The integrated USB and Ethernet interfaces make it easy and reliable to transfer data to peripheral devices or a network.
Interfaces: Bluetooth (optional), Ethernet (LAN), RS232 (integrated/optional), USB-A (to device), USB-B (to device).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty