Would you like to log in again?
Microbalance, XPR3
Supplier: Mettler-Toledo
30875681
77507-144EA 39280 USD InStock
77507-144
Microbalance, XPR3
Balances and Scales
Balance XPR3 gives a supreme performance for micro samples.
  • Precise micro weighing with highest user safety
  • 3.1 g capacity
  • 1 µg readability
  • Integrated methods
  • Results notepad
  • 2 displays
  • Automatic draft shield
  • All-round visibility
  • Small footprint
  • Easy cleaning
  • LabX compatible
Exceptional accuracy for small samples:
With one microgram readability, low minimum weight, and unique design features, you can accurately weigh tiny samples with the utmost safety.
Audit-proof results - Every time:
Innovative quality assurance functions actively monitor the status of your balance and are your reassurance of accurate, repeatable, and valid results.
Small footprint and two terminals:
XPR microbalances have the smallest footprint on the market, plus the main terminal can be placed away from the balance, wherever is most convenient.
Interfaces:
Bluetooth (optional)
Ethernet (LAN)
RS232 (integrated/optional)
USB-A (to device)
USB-B (to device)
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty