Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Human Recombinant LRRK2 (from Baculovirus (Sf9 Insect cells))

Supplier: Sino Biological
L10-12GG-10
77507-240EA 368 USD InStock
77507-240
Human Recombinant LRRK2 (from Baculovirus (Sf9 Insect cells))
Enzymes
Recombinant human LRRK2 (G2019S) (968-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.

  • Gene ID: NM_198578

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty