Would you like to log in again?
Achromobacter Lyticus Recombinant Lys-c (from E. coli)
Supplier: Sino Biological
L585-31HP-500 L585-31HP-20 L585-31HP-01
77507-256EA 1105 USD InStock
77507-256 77507-254 77507-252
Achromobacter Lyticus Recombinant Lys-c (from E. coli)
Enzymes
Recombinant Achromobacter lyticus Lys-c (lysyl-endo-peptidase) (206-473aa) was expressed in E. coli cells using a C-terminal His tag.
The enzyme commission number is EC3.4.21.50.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty