Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Human Recombinant WIP1 (from Baculovirus (Sf9 Insect cells))

Supplier: Sino Biological
P04-20G-10
77507-288EA 328 USD InStock
77507-288
Human Recombinant WIP1 (from Baculovirus (Sf9 Insect cells))
Enzymes
Recombinant full-length human WIP1 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.

  • Gene ID: BC016480

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty