Would you like to log in again?
Human Recombinant PI3K (from Baculovirus (Sf9 Insect cells))
Supplier: Sino Biological
P28-102H-10
77507-312EA 418 USD InStock
77507-312
Human Recombinant PI3K (from Baculovirus (Sf9 Insect cells))
Enzymes
Recombinant full-length human p110 beta and human p85 alpha were co-expressed by baculovirus in Sf9 insect cells using an N-terminal His tag on both proteins.
  • Gene ID: NM_006219, NM_181523
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty