Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Human Recombinant PTPN2 (from E. coli)

Supplier: Sino Biological
P29-20G-10
77507-314EA 248 USD InStock
77507-314
Human Recombinant PTPN2 (from E. coli)
Enzymes
Recombinant full length human PTPN2 was expressed in E. coli cells using a N-terminal GST tag.

  • Gene ID: NM_080422

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty