Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Human Recombinant VCP (from Baculovirus (Sf9 Insect cells))

Supplier: Sino Biological
V116-310G-10
77507-366EA 258 USD InStock
77507-366
Human Recombinant VCP (from Baculovirus (Sf9 Insect cells))
Enzymes
Recombinant full-length human VCP was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.

  • Gene ID: NM_007126

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty