Would you like to log in again?
Universal Pipette Tips
Supplier: Nest Scientific
313001 303023 305052 303001 302023 301023 304052 311012 302106 316001 304023 306023 314012 317012 303206 302012 301052 319012 301012 306012 317001 312012 315001 306001 302001 312001 305012 302052 316012 304012 304001 311001 305023 313012 315012 303012 305001 301001
77507-466CS 217.36 USD InStock
77507-466 77507-488 77507-564 77507-486 77507-464 77507-562 77507-502 77507-546 77507-568 77507-468 77507-500 77507-544 77507-566 77507-506 77507-504 77507-508 77507-490 77507-572 77507-550 77507-494 77507-570 77507-470 77507-510 77507-598 77507-476 77507-498 77507-574 77507-552 77507-474 77507-496 77507-514 77507-558 77507-556 77507-478 77507-518 77507-516 77507-480 77507-484
Universal Pipette Tips
Pipette Tips
NEST aerosol filter pipette tips are designed and manufactured using premium-grade 100% polypropylene for the most critical molecular biology liquid handling applications.
  • Certified RNase, DNase, Human DNA, Pyrogen-Free
  • Low-Retention, Fine-Point design for lowest sample adhesion
  • Ultra-Clear polymer withstands organic solvents and provides clear, error-free pipetting
  • Effective hydrophobic aerosol barrier eliminates carry-over and pipettor contamination
  • Gamma-irradiated Sterile and Graduated
  • Universal-fit for most single and multichannel pipettors
All aerosol filter tips are gamma irradiated sterile and certified RNAse/DNAse-free, human DNA-free, and pyrogen-free. In addition, Nest aerosol filter tips are ultra-clear with a fine-point design for the convenient, precise pipetting of precious nucleic acid and protein samples and are designed to be universal-fit for the majority of research-grade pipettors.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty