Would you like to log in again?
Pipette Tips
Supplier: Nest Scientific
307012 307013 307123 307022 307113 307112 307122 307121 307023 307111 307021 307011
77507-532CS 184.53 USD InStock
77507-532 77507-542 77507-520 77507-530 77507-540 77507-536 77507-524 77507-534 77507-522 77507-528 77507-538 77507-526
Pipette Tips
Pipette Tips
NEST 5 and 10 ml pipette tips are precision-molded from high purity, DNase RNase and pyrogen free, virgin polypropylene.
  • Autoclavable at 121 ℃
  • Meet a wider range of volumetric pipetting needs for more precise pipetting
They are available in 5 and 10 ml, filtered or non-filtered. Choose from compatible pipettors either for Eppendorf, Sartorius, Gilson, or for Thermo pipettors.
They allow pipetting from narrow volume measuring equipment. Clean, hydrophobic surfaces reduce sample retention, and increase accuracy and reproducibility.
Certifications: ISO Class 08.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty