Would you like to log in again?
Storage Bottles, Round
Supplier: Nest Scientific
337301 341101 336201 341301 340201 338101 335201 339301 335101 338301 335301 336101 339201 339101 337201 340301 341201 337101 340101 338201 336301
77507-730CS 142.95 USD InStock
77507-730 77507-740 77507-734 77507-712 77507-722 77507-710 77507-732 77507-742 77507-720 77507-738 77507-716 77507-704 77507-726 77507-736 77507-714 77507-702 77507-724 77507-708 77507-718 77507-706 77507-728
Storage Bottles, Round
Bottles Plastic Bottles
Round Storage Bottles are manufactured in 100,000 grade cleaning room with USP 6 grade raw materials.
  • Excellent physical and chemical indicators
  • Good pressure-resistance
  • Impact-resistance
  • Acid and alkali resistance
  • Autoclave sterilization at 121 ℃, HDPE can be stored at −80 ℃
  • No biotoxicity, pyrogen-free, DNAse/RNAse-free
Sterile and enzyme-free plastic reagent bottles, suitable for product packaging and storage requirements in molecular biology and Cell Biology, laboratory medicine, genomics and proteomics and other areas. Improvement of the quality of product packaging helps to improve the company image and brand value, lets your product packaging meet international standards and enter the international market.
Washing-free, no need for tedious pre-cleaning processing work, ready to use, greatly improves the packaging effect of users. Thickened middle packaging to ensure transportation and storage safety. Use of professional leaking-proof design for bottle mouth, with no need for inner cover or inner gasket for protection, excellent sealability. Guarantee of being 100% leaking-proof to ensure safety even during air transportation, wide-mouthed design for easy access of liquid.
Certifications: ISO 9001&13485 certified.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty