Would you like to log in again?
Automation/Robotic Tips for Tecan and Hamilton Systems
Supplier: Nest Scientific
345112 332112 345269 345013 332013 345212 332002 332216 332102 345012 332202 332003 345102 332217 332113 332103 332107 332016 345169 345113 345215 345205 345115 345105 345002 332116 345005 345069 332117 345015 345202 345213 332203 332017 332012 332212 332207 332213
77507-642CS 197.45 USD InStock
77507-642 77507-664 77507-840 77507-662 77507-844 77507-822 77507-668 77507-644 77507-842 77507-666 77507-826 77507-848 77507-824 77507-846 77507-648 77507-828 77507-670 77507-850 77507-652 77507-674 77507-650 77507-672 77507-656 77507-854 77507-832 77507-678 77507-830 77507-852 77507-654 77507-676 77507-836 77507-856 77507-834 77507-658 77507-838 77507-682 77507-660 77507-680
Automation/Robotic Tips for Tecan and Hamilton Systems
Pipette Tips
NEST automation/robotic tips can be used for research and development and other commonly used applications with needs of high-throughput pipetting in genomics, proteomics, cytomics, immunoassays, metabolomics, biopharmaceuticals.
  • Adopt medical grade polypropylene material, which is manufactured by precision mold, and the material batch is stable
  • DNase-and RNase-free, no heat source contamination
  • Electron beam sterilization
  • Manufacturing facility equipped with class 100,000 clean-room, products packing under sterile condition
  • Unique process technology ensures that the inner surface of the tip is smooth and reduces liquid residue
  • Online testing equipment is used to control the size and concentricity of products in the production line
  • Both conductive and clear are available
Compatible with Hamilton/Tecan/Beckman/Agilent Systems. Filter or non-filter style can be selectable, it has super hydrophilicity, it can shield aerosol, and eliminate the risk of cross-contamination of samples.
Variety of packaging are available. Carry out structural drawing and measurement according to the original adapter to ensure that the product has good air tightness and adaptability.
Effective quality inspections are conducted for strict quality management and control per the client’s needs, including tests for DNase, RNase, proteases and pyrogens.
TECAN automation conductive tips may be used with the TECAN Freedom EVOlyzers and fully automatic sampling systems.
HAMILTON automation conductive tips are used in conjunction with Hamilton series workstations.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty