Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Storage Bottles, Square, Sterile

Supplier: Nest Scientific
352513 354513 334511 354113 334513 354511 333513 352511 354111 353511 355114 333624 333511 353513
77507-972CS 395.76 USD InStock
77507-972 77507-686 77507-696 77507-960 77507-970 77507-954 77507-976 77507-688 77507-698 77507-962 77507-968 77507-956 77507-966 77507-692
Storage Bottles, Square, Sterile
Bottles Cell Culture Bottles
NEST’s Square storage bottle is an ideal option of storage and transportation container for culture medium, serum, buffer, intermediates and reagents for biopharmaceutical and biotechnology companies, reagent R and D institutions, etc.

  • PET wall is light in weight and resistant to low temperature, while PETG wall is thick, durable, and resistant to fracture
  • High chemical durability, suitable for the storage of high purity reagent, standard particle materials and samples
  • High transparency, strong mechanical strength and shock resistance, and convenient for observation and transportation
  • Highly tolerant of low temperature, UV rays, and resistant to fracture
  • Smooth inner surface can minimize the residue on the wall
  • Sterile, DNase/RNase/Pyrogen free

Applications: Storage or sampling of active pharmaceutical ingredients and bulk intermediates, experiments or storage of buffers, culture solutions, intermediates and supernatants, etc, storage of specific PH-sensitive liquid.

Certifications: The two kinds of raw materials of the bottle, PET and PETG, are both in line with USP Class VI Class USP<661> biocompatibility requirements and ISO10993 Quality System.

Caution: Unfit for autoclave. Not a vessel to be heated.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty