Would you like to log in again?
Silicone Sealing Mats
Supplier: Nest Scientific
506002 506062 506016 506061 506102 506005 506064 506066 506001 506004 506063 506006 506162 506003 506065
77508-180CS 227.29 USD InStock
77508-180 77508-190 77508-182 77508-170 77508-192 77508-184 77508-172 77508-164 77508-186 77508-174 77508-166 77508-188 77508-176 77508-168 77508-178
Silicone Sealing Mats
Seals Microplate Seals
The product is easy to open and close, and can effectively seal the deep well plate, which is convenient for operation and saves time.
Good sealing performance can prevent the volatilization of a variety of solvents. Chemical corrosion resistance and is used for low-temperature preservation. The pre-opening design allows the pipette tip to enter the hole without damage to the silicone sealing mat cover.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty