Would you like to log in again?
Sample Vials, with HDPE Caps
Supplier: Nest Scientific
625011 619101 619107 625001 624207 608201 619201 625017 608207 624201 619011 619017 625007
77508-450CS 406.8 USD InStock
77508-450 77508-440 77508-410 77508-442 77508-302 77508-412 77508-444 77508-304 77508-446 77508-448 77508-404 77508-406 77508-408
Sample Vials, with HDPE Caps
Vials
Polypropylene vials, with attached or separated HDPE Caps.
  • Self-standing: the vial placed on the lab workbench steadily even without a base
  • Sterilized by E-beam, Sterility Assurance Level: SAL 10⁻⁶
  • Safe, fast, environment-friendly, no chemical residues
  • DNAse-, RNAse-, pyrogen-,endotoxin-free
  • Disposable
Excellent sealing performance, no leakage of negative pressure 95 Kpa, ensuring safe and effective air transportation.
Temperature range: Approx.–196 to 121 °C
Certifications: Meets USP VI standard.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty