Would you like to log in again?
Storage Bottles, Square, Closed System Solution
Supplier: Nest Scientific
C51121-BHB060A C51022-AAB060A C50921-BBB060A C50922-AGB060A C50921-BHB060A C51022-AGB060A C51121-BBB060A C50921-DZB060A C50922-AAB060A C511AB-AGB060A C51021-DZB060A C51021-BBB060A C51121-DZB060A C51122-AFB060A C51122-AGB060A C51122-AAB060A C51021-BHB060A
77509-152CS 1597.33 USD InStock
77509-152 77509-174 77509-150 77509-160 77509-170 77509-180 77509-128 77509-138 77509-124 77509-168 77509-146 77509-156 77509-134 77509-132 77509-176 77509-164 77509-142
Storage Bottles, Square, Closed System Solution
Bottles Cell Culture Bottles
The closed system storage bottles collection provides an aseptic liquid infusion solution for pharmaceuticals, biotechnology industry and laboratories.
  • Various tubing specifications
  • The injection-molded transfer cap is highly compatible with other major brands
  • The bottle body is made of PETG, which has high transparency, mechanical strength, low-temperature resistance, and UV resistance, making it suitable for observation and transportation
  • The unique external thick collar design of the bottle neck secures a sealed environment for experiments
  • Non-cytotoxic, non-pyrogenic, no animal-derived components
  • Ten bottles per case
  • Pore size: 0.2 µm
  • Membrane are: 4.5 cm²
A standard Luer head is used in the joint of the system, enabling the operation more convenient. Also, the TPE material of tubes is in line with the requirements for industrial pharmaceutical enterprises, and is validated by a certified third party in accordance with BPOG Guide for Evaluating Extractables and Leachables.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty