Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Autofil® Polypropylene Bottle Top Vacuum Filter Funnel Assembly for Solvents

Supplier: Foxx Life Sciences
Autofil®
325-5271-FLS 325-1441-FLS 325-5241-FLS 325-5451-FLS 325-1451-FLS 325-1271-FLS 325-5441-FLS 325-1241-FLS
77509-208CS 156.69 USD InStock
77509-208 77509-206 77509-204 77509-202 77509-212 77509-200 77509-210 77509-198
Autofil® Polypropylene Bottle Top Vacuum Filter Funnel Assembly for Solvents
Filters Bottle Top Filters
Polypropylene funnel assembly for solvents with a GL45 neck, three menbrane types, Nylon, PES and PTFE.

  • Non sterile
  • Bottle top holders allow hands-free operation of the filter unit
  • Funnel height: 109 mm (4.3")
  • Diameter (max): 114 mm (4.5")
  • Choice of funnel volume: 500 or 1000 ml
  • Funnel material: Polypropylene
  • Filter material: PTFE, Nylon or PES
  • Filter pore size: 0.2 or 0.45 μm

Foxx Life Sciences' Autofil® PP bottle-top filters for solvent filtration are a revolutionary way to purify solvents for analytical applications to obtain sterilized or purified extracts. Foxx has developed these single piece polypropylene funnel, designed to withstand harsh solvents that would normally degrade traditional bottle-top filter assemblies. All active components are FDA-grade which makes Autofil® PP perfect for the extraction of compounds used in the food and drug industry.

Certifications: In compliance with ISO 13485 standards.

Packaging: The bottle top filters are shrinked-wrapped.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty