Would you like to log in again?
RingSeal™ Cryogenic Vials
Supplier: Globe Scientific
RingSeal™
3034-5 3034-2-RB 3033-CIB 3034-1 3033-CIR 3034-2 3033-CIW 3034-4 3032-1 3032-5 3034-5-RB 3032-2-RB 3033-CIG 3032-5-RB 3033-CIY 3032-4 3032-2
77509-262CS 175.77 USD InStock
77509-262 77509-240 77509-250 77509-260 77509-238 77509-248 77509-258 77509-236 77509-246 77509-256 77509-234 77509-244 77509-254 77509-232 77509-264 77509-242 77509-252
RingSeal™ Cryogenic Vials
Vials
A good choice for long-term sample storage. Built with reliable O-ring seals, this exceptional range offers multiple variants to meet diverse research needs.
  • RingSeal™ Cryogenic Vials provide leak-proof protection up to 90 kPa, safeguarding against potential leaks or contamination
  • Reliable O-ring seals effectively protect samples within a wide temperature range, from −196 to 121 °C
  • Choose between round bottom and self-standing vials, based on specific requirements
  • Certified DNase-, RNase-, and Pyrogen-free vials ensure the integrity and cleanliness of valuable samples
  • Available internal and external thread options allow compatibility with a variety of laboratory processes
  • Large writing area aids in convenient sample identification and organization
  • Autoclavable vials ensure efficient sterilization while maintaining structural integrity
Globe Scientific’s RingSeal™ Cryogenic vials are available in both internal and external thread options, as well as self-standing and round bottom formats. Vials are offered in the most popular volumes.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty