Would you like to log in again?
Durx® 670 AS (Aerospace) Non-woven Cleanroom Wiper, 55% Cellulose/45% Polyester
Supplier: Berkshire
DurxDurx® 670 AS (Aerospace)
DR670121220AS DR670090920AS
77528-884CS 479.17 USD InStock
77528-884 77528-876
Durx® 670 AS (Aerospace) Non-woven Cleanroom Wiper, 55% Cellulose/45% Polyester
Wipes Cleanroom Wipes Dry Cleanroom Wipes
Durx® 670 AS for Aerospace (AS) is a hydroentangled, non-woven blend of 55% cellulose and 45% polyester with no binders or other chemical additives. This wiper meets the AMS 3819D, Class 2, Grade A, Form 1 requirements.
  • Excellent combination of the synthetic polyester strength and cleanliness, with the absorbent characteristics of cellulose
  • Smooth, highly absorbent, and durable
  • Low extractable levels and particle counts
  • Chemically compatible with common cleaning solutions
  • Economical
Durx® 670 AS for Aerospace (AS) is a hydroentangled nonwoven blend of 55% cellulose and 45% polyester with no binders or other chemical additives and meets the AMS 3819D, Class 2, Grade A, Form 1 requirements. This combination of properties provides Durx® 670 AS with the absorbency level, abrasion resistance, and chemical compatibility required for controlled environments and applications where both cleanliness and economy are critical. Durx® 670 AS offers application versatility to meet the specific challenges faced in aerospace processes and applications.
Certifications: Meets AMS 3819D requirements, Class 2, Grade A, Form 1. Meets BMS 15-5 requirements. ISO Class 5 to 8, Cleanroom Class 100 to 100,000. EU Grade B/C/D.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty