Would you like to log in again?
MicroSeal SuperSorb® AS (Aerospace) Sealed Edge Cleanroom Laundered Knit Wiper, 100% Polyester
Supplier: Berkshire
MicroSeal SuperSorb® AS (Aerospace)
MSSS121212AS MSSS09098AS
77528-880CS 923.61 USD InStock
77528-880 77528-878
MicroSeal SuperSorb® AS (Aerospace) Sealed Edge Cleanroom Laundered Knit Wiper, 100% Polyester
Wipes Cleanroom Wipes Dry Cleanroom Wipes
MicroSeal SuperSorb® AS for Aerospace is a high sorbency, ultrasonically sealed edge cleanroom laundered two-ply wiper that meets AMS 3819D, Class 4, Grade A, Form 1 requirements.
  • Critically low particles, fibers, ions, and extractables
  • High abrasion resistance
  • Chemically compatible with IPA, Acetone, and other solvents
  • Highest sorbency level available in a polyester knit reduces wiper usage
  • Pinsonic design adds surface texture that aids in stubborn residue removal
MicroSeal SuperSorb® AS for Aerospace is a high sorbency ultrasonically sealed edge cleanroom laundered two-ply wiper that meets AMS 3819D, Class 4, Grade A, Form 1 requirements. It is composed of 100% continuous filament polyester knit fabric and has high cleanliness levels, abrasion resistance, and chemical compatibility. These properties make it ideal for ISO Class 3 and above environments and applications requiring critical contaminant control combined with superior performance. MicroSeal SuperSorb® AS provides the highest level of sorbency available in a two-ply polyester knit application and offers versatility to meet the specific challenges faced in aerospace processes and applications.
Certifications: Meets AMS 3819D requirements, Class 4, Grade A, Form 1. Meets BMS 15-5 requirements. ISO Class 3 to 5, Cleanroom Class 1 to 100. EU Grade B/C/D.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty