Would you like to log in again?
Sol-M® Luer Adapter
Supplier: Sol-Millennium
Sol-M®
LA21920
77528-890CS 327.33 USD InStock
77528-890
Sol-M® Luer Adapter
Adapters Adapters, Joints with Tube Connectors
The Sol-M® luer adapter is designed to work in a variety of blood collection applications by securely connecting to female Luer connections.
  • Blue cap to differentiate from other blood collection devices
  • Designed to ensure secure connections
An ideal accessory to use with blood collection sets that feature a holder for evacuated tubes. Fitted with rubber sleeve to ensure no blood spills or exposure.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty