Would you like to log in again?
AlphaTec® 53-003 Broad Chemical Protection Gloves with Nylon Liner
Supplier: Ansell
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
AlphaTec®
53003100 53003080 53003070 53003110 53003090
77528-914CS 1617.31 USD InStock
77528-914 77528-928 77528-906 77528-896 77528-922
AlphaTec® 53-003 Broad Chemical Protection Gloves with Nylon Liner
Gloves
Revolutionary Hybrid Technology delivering a breakthrough in chemical performance.
  • MICROCHEM™ multi-layer barrier features unique rubber polymer and plastic laminate technology
  • Designed to provide extreme protection against a wide range of chemical types
  • Contact heat approved up to 100 °C/212 °F
  • Anti-static according to EN 1149-3 standards
  • Made without natural rubber or silicone, ensuring protection against mechanical risks and allergic reactions
  • Unique manufacturing process and nylon liner to enhance comfort during use
The AlphaTec® 53-003 series are revolutionary industrial gloves that redefine chemical protection. The innovative design combines an advanced rubber polymer coating and plastic laminate technology improving chemical permeation while delivering comfort and tactility. These safety gloves meet the stringent requirements of EN ISO 374:2016, with 17 out of 18 candidate chemicals achieving a minimum Class 2 classification, and 16 out of 17 achieving class 6>480 minutes. In addition to chemical protection, they satisfy EN 407 contact heat level 1, preventing workplace injuries. They also possess EN-compliant abrasion resistance.
The unique manufacturing process allows for the glove to be form fitting, offering elevated comfort and dexterity.
Certifications: CE 0493; EN 421:2010; EN ISO 374-5:2016; EN 388:2016+A1:2018; EN407:2020.
Packaging: 6 pairs per bag, 12 bags per carton.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty