Would you like to log in again?
EDGE® 48-707 Medium-Duty Cut Protection Industrial Gloves
Supplier: Ansell
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
EDGE®
48707090 48707100 48707070 48707110 48707080 48707060
77528-904CS 753.31 USD InStock
77528-904 77528-902 77528-934 77528-918 77528-894 77528-910
EDGE® 48-707 Medium-Duty Cut Protection Industrial Gloves
Gloves
Affordable safety gloves, offering superb grip as well as cut and abrasion resistance.
  • Very high cut protection for robust mechanical protection gloves
  • Good grip and dexterity provide secure handling for precision tasks
  • Anti-static design reduces the risk of electrostatic shock
Advanced protection EDGE® 48-707 cut- and abrasion-resistant gloves provide ANSI cut level A6 and EN 388 ISO cut level F, which afford a very high level of cut protection. Owing to their palm PU coating, these protective gloves combine good grip as well as dexterity, and are suitable for use in dry or slightly oily environments. In addition, EDGE® 48-707 ESD gloves meet the requirements for electrostatic dissipative materials as per EN 1149. These grey hand protection gloves also possess a seamless liner and knitted wrist, which deliver a secure fit for use in applications that require high cut resistance.
Certifications: ANSI ABR 5; ANSI Cut A6; ANSI/ISEA 105-2016 PUN 4; Ecovadis label; EN 388:2016+A1:2018; EN ISO 21420:2020.
Packaging: 12 pairs per bag, 144 pairs per carton.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty