Would you like to log in again?
HyFlex® 11-281 Comfortable Sleeves with INTERCEPT™ Technology
Supplier: Ansell
HyFlex®
11281220WVEL
77528-930BX 378.46 USD InStock
77528-930
HyFlex® 11-281 Comfortable Sleeves with INTERCEPT™ Technology
Gloves
Protective arm sleeves with thumb slot and VELCRO, combining cut protection with unmatched comfort.
  • INTERCEPT™ cut resistance; innovative knitwork for advanced defense
  • HPPE yarn offers protection with cool, breathable comfort
  • Varied sizing ensures a perfect fit, with various width and length options
INTERCEPT™ cut resistance technology's mix of natural and synthetic fibers guarantees ANSI A4 and EN ISO D cut protection, the perfect pairing for cut-resistant HyFlex® gloves. The HyFlex® 11-281 protective sleeves HPPE yarn offers wearers cool, breathable all-day comfort, with a thumb slot to keep them firmly in place. With multiple width and length options, there is a HyFlex® 11-281 sleeve to fit almost every worker.
HyFlex® 11-281 protective sleeves also comply with FDA regulations 21 CFR 177.1520 and 21 CFR 177.2600 for safe food handling. They possess Oeko-Tex® certification, indicating they are free of substances harmful to human health.
Certifications: CE Number; ANSI Cut A4; Ecovadis label; EN 388:2016+A1:2018; EN ISO 21420:2020; Food Contact; OEKO-TEX® CITEVE; Category II; REACH Compliant.
Packaging: 50 pieces per box.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty