Would you like to log in again?
AlphaTec® 53-002 Broad Chemical Protection Gloves
Supplier: Ansell
AlphaTec®
53002110 53002090 53002100 53002080 53002070
77528-924CS 1174.18 USD InStock
77528-924 77528-916 77528-938 77528-932 77528-898
AlphaTec® 53-002 Broad Chemical Protection Gloves
Gloves
Revolutionary hybrid technology delivering a breakthrough in chemical performance.
  • MICROCHEM™ multi-layer barrier features unique rubber polymer and plastic laminate technology
  • Designed to provide extreme protection against a wide range of chemical types
  • Anti-static according to EN 1149-3 standards
  • Made without natural rubber or silicone, ensuring protection against mechanical risks and allergic reactions
  • Unique donning and doffing layer to allow ease of use
The AlphaTec® 53-002 series are revolutionary industrial gloves that redefine chemical protection. The innovative design combines an advanced rubber polymer coating and plastic laminate technology improving chemical permeation while delivering comfort and tactility. These safety gloves meet the stringent requirements of EN ISO 374:2016, with 17 out of 18 candidate chemicals achieving minimum Class 2 classification, and 16 out of 17 achieving class 6>480 minutes.
The unique manufacturing process allows for the glove to be form fitting, offering elevated comfort and dexterity.
Certifications: CE 0493; EN ISO 374-5:2016; EN 388:2016+A1:2018; EN 421:2010; EN ISO 374-1:2016.
Packaging: 6 pairs per bag, 12 bags per carton.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty