Would you like to log in again?
Teknova Complete Minimal Dropout Plates Glucose with a Single Dropout
Supplier: Teknova
C3040 C3061 C3080 C3040 C3000 F2344 C3021 C3041 C3061 C3060 C3000 C3041 C3060 F2344 C3020 C3060 C3020 C3081 C3061
77443-464EA 82.38 USD InStock
77443-464 100217-532 100217-540 CA100217-826 CA100217-822 CA100217-536 CA100217-538 100218-978 CA100217-824 CA100217-532 CA100217-544 77528-946 CA100217-832 CA100217-540 CA100218-978 100217-538 100217-824 100217-826 100217-536
Teknova Complete Minimal Dropout Plates Glucose with a Single Dropout
Media for Microbiology Prepared Media for Microbiology
Plates are shelf-life dated and guaranteed for 4 weeks from the date of shipment
All plates must be stored at 4°C and in the absence of continuous exposure to light. With proper storage, most antibiotic plates are useable for up to 2 months. Complete Minimal Dropout Plates (CM)Also known as Synthictic Dropout Plates (SD). All plates sold in sleeves of 10.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty