Would you like to log in again?
LB Agar (Lennox), Teknova
Supplier: Teknova
L3500 L3500
101320-088PK 171.84 USD InStock
101320-088
LB Agar (Lennox), Teknova
Media for Microbiology Prepared Media for Microbiology
LB Lennox is used for the growth of recombinant Escherichia coli and other enteric bacteria
  • Growth of recombinant Escherichia coli
  • Commonly used for cloning
  • Selection with salt-sensitive antibiotics
This LB agar plate contains 1% tryptone, 0.5% yeast extract, 0.5% Nacl, 1.5% agar.
This is commonly used for cloning in recombinant E. coli and selection with salt-sensitive antibiotics such as Blasticidin, CloNat, Hygromycin, Puromycin and Zeocin.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty