Would you like to log in again?
LB Agar Plates with 100 µg/ml Kanamycin, Teknova
Supplier: Teknova
L5027 L1027 L3027 L3027 L1027 L5027 L5027 L1027
77443-344EA 82.38 USD InStock
77443-344 CA100217-258 100217-258 77528-980 100218-234 CA100218-234 100216-592 CA100216-592
LB Agar Plates with 100 µg/ml Kanamycin, Teknova
Media for Microbiology Prepared Media for Microbiology
Ready to use LB agar plates (Miller formulation) for the growth of recombinant E. coli. All plates are made with the highest quality products and tested with four strains of E. coli.
Ingredients: 1.0% tryptone, 0.5% yeast extract, 1.0% sodium chloride, 1.5% agar, 100 µg/ml kanamycin.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty