Would you like to log in again?
LB Agar Plates with 150 µg/mL Carbenicillin, Teknova
Supplier: Teknova
L6011 L6011 L6011
200066-622PK 245.9 USD InStock
CA200066-622 200066-622 77528-984
LB Agar Plates with 150 µg/mL Carbenicillin, Teknova
Media for Microbiology Prepared Media for Microbiology
Vented Qtray bioassay tray.
  • 4 pack
  • Sterile
Contains 1% Tryptone, 0.5% yeast extract, 1.0% sodium chloride, 1.5% agar, 150 µg/mL carbenicillin.
Ambient shipping conditions. Refrigerate to store.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty