Would you like to log in again?
LB Agar Plates, Teknova
Supplier: Teknova
L1100 L5100 L5100 L2100-4PK L2100 L7200 L1100 L7200 L3100 L3100 L1100 L7200
100218-472PK 1268.95 USD InStock
CA100220-120 100218-472 100218-252 CA100218-472 CA100218-252 77443-316 100217-280 77528-992 100216-614 CA100217-280 100220-120 CA100216-614
LB Agar Plates, Teknova
Media for Microbiology Prepared Media for Microbiology
For the growth of E. coli and other enteric bacteria.
  • Sleeve of 20 plates
  • Sterile
Ingredients: 1.0% Tryptone (10.0 g/L), 0.5% Yeast Extract (5.0 g/L), 1.0% Sodium chloride (NaCl) (10.0 g/L), 1.5% Agar (15.0 g/L).
Ambient shipping conditions. Store at room temperature.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty