Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tryptic Soy Broth, Teknova

Supplier: Teknova
T1516 T1515 T1516 T9400 T1515 T1550 T9400 T1516 T1570 T1550
101320-664PK 177.87 USD InStock
101320-664 CA100220-230 CA101320-664 101320-708 CA101320-708 77529-012 101447-050 100218-958 100220-230 CA100218-958
Tryptic Soy Broth, Teknova
Media for Microbiology Prepared Media for Microbiology
Tryptic Soy Broth is a general purpose media for growth of bacteria that have high nutritional requirements.

  • Ambient shipping conditions
  • Store at room temperature

Contains 1.7% tryptone, 0.3% soytone, 0.25% Glucose, 0.5% sodium chloride, and 0.25% potassium phosphate.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty